missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16246-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
SAT2 is an enzyme which catalyzes the acetylation of polyamines. Substrate specificity: norspermidine > spermidine = spermine >> N(1)acetylspermine = putrescine.Especificaciones
SAT2 | |
Polyclonal | |
Unconjugated | |
SAT2 | |
Diamine acetyltransferase 2, SAT2, SSAT2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
112483, 360547, 69215 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6P8J2, Q96F10 | |
Sat2 | |
SAT2 Fusion Protein Ag9179 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |