missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Sartoclear Dynamics™: Lab V

For simultaneous clarification and sterile filtration of 50 mL to 1 L mammalian cell cultures in one step without centrifugation.

Marca:  Sartorius™ SDLV-1000-10C--2

Detalles adicionales : Peso : 1.36000kg

 Ver más versiones de este producto

Código de producto. 15935840

  • 937.58€ / paquete de 24


Consúltenos : 902 239 303 
Añadir a la cesta

Descripción

Descripción

Sartoclear Dynamics™ Lab V kits are designed for simultaneous clarification and sterile filtration of 50 mL to 1 L of mammalian cell culture broth to prepare samples for subsequent downstream processing steps, such as protein purification and concentration. The convenient kits combine pouches of filter aid for clarification and vacuum filtration units for sterile filtration.

The Sartoclear Dynamics™ Lab V Kits combine the use of a filter aid for clarification and a vacuum filtration unit with a 0.22 µm polyethersulfone membrane for sterile filtration of up to 1 L mammalian cell cultures. The ready-to-use filter aid is supplied in dust-free and gamma-irradiated pouches.

The Sartolab™ RF bottletop filters with receiver flasks, sterilized by gamma irradiation are
available in volume capacities ranging from 150 mL to 1,000 mL.

  • Clarify high-density mammalian cell culture in minutes
  • Out-of-the-box solution
  • Fast and effortless
  • No need for centrifugation

Ten kits are available to meet all your needs based on your
filtration volume and the cell density of your mammalian
cell cultures.

Especificaciones

Especificaciones

Diatomaceous Earth/Heavy metal-free polystyrene/Heavy metal-free polyethylene
Sartoclear Dynamics&trade: Lab V kits have been designed for cell harvesting of mammalian cell cultures, such as CHO, HEK, hybridomas and many others, with high cell densities.
mammalian cell harvesting/culture
Each Sartoclear Dynamics&trade: Lab V kit is comprised of pouches of filter aid and vacuum filtration units (Sartolab&trade: RF) that match your needs. Find the right kit in just two easy steps: 1. Determine the volume range of your sample to be filtered. 2. Then reference this range to the cell density of your cell culture
10g
1,000mL
24 x 1L Filter Units + 10g Filter Aid
0.22 μm polyethersulfone/high flow rate
ambient
2 × 180C3----------E (24 × Sartolab&trade: RF 1000; 0.22 μm: PES; single-packaged; sterile), 1 × SDLKG-10.0------2 (24 × pouches of filter aid, 10 g)
Videos
Documentos