missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Clorhidrato de sarpogrelato 98 %
Click to view available options
Cantidad:
250 mg
1 g
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Nombre del producto químico o material | Sarpogrelate hydrochloride |
Punto de fusión | 145°C to 148°C |
Intervalo de porcentaje de ensayo | 97.5% min. (HPLC) |
Cantidad | 250 mg |
Información de solubilidad | Solubility in water: soluble. Other solubilities: soluble in dimethylsulfoxide and methanol |
Formula Weight (peso de la fórmula) | 465.97 |
Porcentaje de pureza | 98% |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'