missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAPK4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10217-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a member of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. This kinase is closely related to p38 MAP kinase, both of which can be activated by proinflammatory cytokines and cellular stress. MAP kinase kinases 3, and 6 can phosphorylate and activate this kinase. Transcription factor ATF2, and microtubule dynamics regulator stathmin have been shown to be the substrates of this kinase.Especificaciones
SAPK4 | |
Polyclonal | |
Unconjugated | |
MAPK13 | |
MAP kinase 13, MAP kinase p38 delta, MAPK 13, MAPK13, p38delta, PRKM13, SAPK4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26415, 29513, 5603 | |
-20°C | |
Liquid |
Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O15264, Q9WTY9, Q9Z1B7 | |
MAPK13 | |
SAPK4 Fusion Protein Ag0255 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |