Learn More
SAPAP4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA562919
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: GVQVEDDWRS SVPSHSMSSR RDTDSDTQDA NDSSCKSSER SLPDCTPHPN SISIDAGPRQ APKIAQIKRN LSYGDNSDPA L Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 98%.
SAP90/PSD-95-associated protein 4 (SAPAP4, also known as DLGAP4) is a member of a protein family whose members specifically interact with PSD-95/SAP90, a membrane-associated guanylate kinase localized at postsynaptic density (PSD) in neuronal cells. Like the other SAPAP proteins, SAPAP4 is thought to be an adaptor protein that also interacts with different synaptic scaffolding proteins, cytoskeletal and signaling components, such as focal adhesion kinase (FAK) and proline-rich tyrosine kinase 2 (PYK2). SAPAP4 mRNA is targeted to cell bodies in a similar manner to SAPAP1 and -2, whereas SAPAP3 mRNA is detected mainly in cell bodies.Especificaciones
SAPAP4 | |
Polyclonal | |
Unconjugated | |
DLGAP4 | |
AI225853; BC024558; Dap4; DAP-4; discs large homolog associated protein 4; discs, large (Drosophila) homolog-associated protein 4; discs, large homolog-associated protein 4; discs, large homolog-associated protein 4 (Drosophila); disks large-associated protein 4; DLG associated protein 4; Dlgap4; DLP4; Kiaa0964; PSD-95/SAP90-associated protein-4; PSD-95/SAP90-binding protein 4; RP5-977B1.6; SAP90/PSD-95-associated protein 4; Sapap4; SAPAP-4; WBP16; WW domain-binding protein 16 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22839 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2H0 | |
DLGAP4 | |
Recombinant protein corresponding to Human SAPAP4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only