Learn More
Uvinul A Plus, TRC
Product Code. 17988414
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17988414 5 g 5g
17989844 100 mg 100mg
17908234 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17988414 Supplier Toronto Research Chemicals Supplier No. TRCU8503005G
 Restricted Item
Fecha estimada de envoi: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 302776-68-7
Molecular Formula C24 H31 N O4
Molecular Weight (g/mol) 397.51
Synonym 2-(4'-Diethylamino-2'-hydroxybenzoyl)benzoic acid hexyl ester, Hexyl 2-[4-(diethylamino)-2-hydroxybenzoyl]benzoate, Uvinar A PLUS GRRANULAR, Uvinul A Plus, Uvinul A Plus Granular, Uvinul A+, n-Hexyl 2-(4-diethylamino-2-hydroxybenzoyl)benzoate
IUPAC Name hexyl 2-[4-(diethylamino)-2-hydroxybenzoyl]benzoate
SMILES CCCCCCOC(=O)c1ccccc1C(=O)c2ccc(cc2O)N(CC)CC
Specifications
Quantity 5 g
InChI Formula InChI=1S/C24H31NO4/c1-4-7-8-11-16-29-24(28)20-13-10-9-12-19(20)23(27)21-15-14-18(17-22(21)26)25(5-2)6-3/h9-10,12-15,17,26H,4-8,11,16H2,1-3H3
Formula Weight 397.2253
Percent Purity >95%
Chemical Name or Material Diethylaminohydroxybenzoyl hexyl benzoate
Grade HPLC