Learn More
Thermo Scientific™ Sample Cuvettes for Eutech™ TB 1000 Turbidity Meter
Product Code. 12911778
Change view
Click to view available options
For Use With (Equipment):
CyberScan TB 1000
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
12911778 CyberScan TB 1000 Each
1 options
This item is not returnable. View return policy
Product Code. 12911778 Supplier Thermo Scientific™ Supplier No. 01X265701
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
The CyberScan TB1000 series turbidity meters and cuvettes are designed to meet the stringent requirements of today's scientists.
Sample Cuvettes for Eutech™ TB 1000 Turbidity Meter are borosilicate glass vials with light-shield screwcaps, fill line and indexing marks.The cuvettes come in packs of three.
Features:
  • Pack of 3 sample cuvettes for TB1000W (white light turibidity meter) & TB1000IR( infra-red turbidity meter)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) CyberScan TB 1000
Description Sample Cuvettes
Quantity 3 Pack