Learn More
Lonza™ SAGM™ Small Airway Epithelial Cell Growth Medium SingleQuots™ Supplements and Growth Factors
Product Code. 11645490
Change view
Click to view available options
Quantity:
BPE: 2.0 mL, Insulin: 0.5 mL, Hydrocortisone: 0.5 mL, GA-1000: 0.5 mL, Retinoic Acid: 0.5 mL, Fatty Acid Free BSA: 5 mL, Transferrin: 0.5 mL, Triiodothyronine: 0.5 mL, Epinephrine: 0.5 mL, hEGF: 0.5 mL
Unit Size:
Each
Product Code. Quantity Unit Size
11645490 BPE: 2.0 mL, Insulin: 0.5 mL, Hydrocortisone: 0.5 mL, GA-1000: 0.5 mL, Retinoic Acid: 0.5 mL, Fatty Acid Free BSA: 5 mL, Transferrin: 0.5 mL, Triiodothyronine: 0.5 mL, Epinephrine: 0.5 mL, hEGF: 0.5 mL Each
1 options
This item is not returnable. View return policy
Product Code. 11645490 Supplier Lonza™ Supplier No. CC4124
  • ONLINE EXCLUSIVE PRICE
    366.00€
    285.95€ / paquete de 1
    Ahorro 80.05€
    21% Descuento
 Restricted Item
Fecha estimada de envoi: 12-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Supplements and growth factors kit for use with small airway epithelial cell basal medium.
  • Separately sold SingleQuot™ kit for supplementing basal medium.
  • Part of the full SAGM™ Small Airway Epithelial Cell Growth Medium BulletKit™ (catalog number CC-3118) which consists of basal medium and supplements packaged together.
  • Serum-free and optimized to grow human small airway epithelial cells (SAEC).
  • Includes the following:
    • 1 x Orange Cap Vial with BPE 2.0 mL
    • 1 x Lilac Cap Vial with Insulin 0.5 mL
    • 1 x Natural Cap Vial with Hydrocortisone 0.5 mL
    • 1 x Red Cap Vial with GA-1000 0.5 mL
    • 1 x Amber Vial with Retinoic Acid 0.5 mL
    • 1 x Bottle with Fatty Acid Free BSA 5 mL
    • 1 x Natural Cap Vial with Transferrin 0.5 mL
    • 1 x Amber Vial with Triiodothyronine 0.5 mL
    • 1 x Amber Vial with Epinephrine 0.5 mL
    • 1 x Green Cap Vial with hEGF 0.5 mL
TRUSTED_SUSTAINABILITY
Specifications
Cell Type Airway Cells
Product Type Supplements and growth factors
Quantity BPE: 2.0 mL, Insulin: 0.5 mL, Hydrocortisone: 0.5 mL, GA-1000: 0.5 mL, Retinoic Acid: 0.5 mL, Fatty Acid Free BSA: 5 mL, Transferrin: 0.5 mL, Triiodothyronine: 0.5 mL, Epinephrine: 0.5 mL, hEGF: 0.5 mL
Content And Storage BPE, Insulin, Hydrocortisone, GA-1000, Retinoic Acid, Fatty Acid Free BSA, Transferrin, Triiodothyronine, Epinephrine, hEGF