missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Safranina T, Honeywell™ Fluka™
Click to view available options
Cantidad:
25 g
100 g
Envase:
Frasco de vidrio
Glass Bottle
Descripción
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Especificaciones
Nombre del producto químico o material | Safranin T |
Número UN | NONH for all modes of transport |
Beilstein | 3924099 |
Nombre IUPAC | 2,7-diamino-1,8-dimethyl-5-phenyl-5λ⁵-phenazin-5-ylium chloride |
Formula Weight (peso de la fórmula) | 350.84g/mol |
Envase | Glass Bottle |
Cantidad | 100 g |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'