Learn More
SCAT SafetyWasteCap S60/61 Thread
Safely connect your system waste tubes directly to your disposal system container via the ports in this cap.
Marca:  SCAT 108964
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 10797260
  • 322.10€ / cada uno
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • Variety of waste cap styles available with different connection arrays available
  • Designed to provide fittings and accessories for safe waste collection
  • For containers with S60/61 threads
Especificaciones
Especificaciones
Includes 2 x 2.3/3.2 mm OD capillary connectors, 1 x connector for exhaust filter, 1 x mechanical level control, 1 x ground connection
Waste disposal system
Waste disposal container with S60/61 thread opening
Waste Disposal System Cap