Learn More
Pacific Handy Cutter Safety Point Standard Utility Blade Dispenser
Product Code. 17789027
Change view
Click to view available options
Product Type:
Blade Dispenser
Unit Size:
Pack of 6
Product Code. Product Type Unit Size
17789027 Blade Dispenser Pack of 6
1 options
This item is not returnable. View return policy
Product Code. 17789027 Supplier Pacific Handy Cutter Supplier No. QBD379
  • ONLINE EXCLUSIVE PRICE
    255.60€
    209.78€ / paquete de 6
    Ahorro 45.82€
    17% Descuento
Fecha estimada de envoi: 16-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The QBD-379 Blade Dispenser safely dispenses one SPS-92 Standard Utility Blade at a time by easily pulling the yellow tab. The SPS-92 Blade fits most standard utility knives and features Safety Point Tips.
  • The blunted safety-points help prevent puncture wounds and injuries.
  • The convenient gravity fed dispenser helps control blade inventory and insures that employees have the proper tools where and when needed to remain productive.
  • The dispenser is made of durable plastic and contains 75 new blades.
  • Plus, by pairing a PHC Blade Dispenser (SPD-017, QBD-324, or QBD-379) to the front of the PHC Blade Bank you create a blade change station that will increase the safety and efficiency of your staff.
  • By using both items together, your employees will be encouraged to properly dispose of loose blades and easily access new replacement blades near their work stations and throughout your facility, ultimately increasing overall productivity.
Specifications
Product Type Blade Dispenser
For Use With (Equipment) Easily Access New Replacement Blade
Disposable Single-use
Sterility Non-sterile