Learn More
Euro Protection™ Safety Helmet
Product Code. 12007876
Change view
Click to view available options
Size:
Adjustable (55 to 62 cm)
Unit Size:
Pack of 48
Product Code. Size Unit Size
12007876 Adjustable (55 to 62 cm) Pack of 48
1 options
Product Code. 12007876 Supplier Euro Protection™ Supplier No. 65300
  • ONLINE EXCLUSIVE PRICE
    676.28€
    368.86€ / paquete de 48
    Ahorro 307.42€
    45% Descuento
 Restricted Item
En stock
Add to basket
Edge
Add to basket
Euro Protection™ Safety Helmet is adjustable and constructed of solid polypropylene.
  • Polypropylene construction
  • Sweatband
  • Adjustable fitting by ratchet
Specifications
Certifications/Compliance EN 397
Color White
Material Polypropylene
Size Adjustable (55 to 62 cm)
Weight (Metric) 350 g