Learn More
Asecos™ K-UB-90 Combi Safety Storage Cabinet
Safe and approved storage of aggressive and flammable hazardous substances in work areas.
Marca:  Asecos™ 30358-001-30360
Detalles adicionales : Peso : 162.00000kg
 Ver más versiones de este producto
Código de producto. 15291025
  • 1950.00€ / cada uno
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • Fits under Fume cupboards
  • Robust construction and longevity
  • User friendly and safe
  • Underbench cabinet
  • With cylinder locking and locking state indicator
  • In light grey RAL 7035
  • Sheet steel powder coated RAL 7035
  • Fire resistance of 90 minutes
  • In accordance with EN 14470-1 and TRGS 510
Servicios para Instrumentación y Equipamiento
Five years
Especificaciones
Especificaciones
CE, EN14727
Light Grey (RAL 7035)
Winged
601mm
K90.060.110.050.UB.ST
1102mm
Steel
502mm
Left: 381 x 470 x 502mm, Right: 468 x 440 x 562mm
Storage of flammable liquids
1 x drawer, 2 x pull out shelf
Combi Underbench Cabinet
Gray
Videos