Learn More
SCAT HPLC Safety Caps
Product Code. 16554170 Shop All SCAT Europe Products
Change view
Click to view available options
Thread Size:
B83
GL38
GLS40
GLS80
NS29/32
Includes:
1 PFA Fitting (3.2 mm OD), Air Valve
2 PFA Fittings (3.2 mm OD), Air Valve
3 PFA Fittings (1.6/2.3/3.2 mm OD), 2 PFA Blind Plugs, Air Valve
3 PFA Fittings (3.2 mm OD), Air Valve
4 PFA Fittings (3.2 mm OD), Air Valve
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size Includes Unit Size
16554170 GLS40 3 PFA Fittings (3.2 mm OD), Air Valve Each
10851421 GLS80 1 PFA Fitting (3.2 mm OD), Air Valve Each
10348149 NS29/32 2 PFA Fittings (3.2 mm OD), Air Valve Each
10541728 GLS80 2 PFA Fittings (3.2 mm OD), Air Valve Each
15168299 GL38 3 PFA Fittings (1.6/2.3/3.2 mm OD), 2 PFA Blind Plugs, Air Valve Each
10839170 GLS80 3 PFA Fittings (3.2 mm OD), Air Valve Each
10222440 B83 4 PFA Fittings (3.2 mm OD), Air Valve Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 16554170 Supplier SCAT Supplier No. 107742
  • ONLINE EXCLUSIVE PRICE
    148.00€
    110.64€ / paquete de 1
    Ahorro 37.36€
    25% Descuento
Fecha estimada de envoi: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
These safety caps provide maximum safety for your HPLC solvent supply. Extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapors. Complete with integrated air valve for pressure equalization.
  • High quality materials ensure maximum safety for lab and staff
  • 360° rotatable and up to 200°C autoclavable screw cap for maximum flexibility
  • Stable construction for highest safety and easy handling
  • Chemically resistant against aggressive organic solvents
  • Attractive design and clear arrangement of the connections
  • Improved design for easy connection of capillaries in various colors
  • Free air valve with change indicator included with every SafetyCap
Specifications
For Use With (Equipment) Lab Bottles
Includes 3 PFA Fittings (3.2 mm OD), Air Valve
Material PTFE
Thread Size GLS40
For Use With (Application) HPLC Systems
Autoclavable Autoclavable
Product Type Safety Cap III