Learn More
Justrite™ Safety Cabinet Slimline
Product Code. 11975426
Change view
Click to view available options
Door Style:
Self-close
Unit Size:
Each
Product Code. Door Style Unit Size
11975426 Self-close Each
1 options
This item is not returnable. View return policy
Product Code. 11975426 Supplier Justrite™ Supplier No. 8922221
  • ONLINE EXCLUSIVE PRICE
    2790.03€
    2110.63€ / paquete de 1
    Ahorro 679.40€
    24% Descuento
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
1 door, self close, not for phenol, nitric /sulphuric acid, steel, double wall, epoxy powder finish, dual vents, earth wire connections, leakproof sills, levelling feet blue 1,651mm x 591mm x 457mm (w x h x d) 83L
Specifications
Door Style Self-close