missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100B Mouse, Clone: 4C4.9 + S100B/1012, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 6285-MSM3-P0
Detalles adicionales : Peso : 0.01000kg
Especificaciones
S100B | |
Cytoplasmic | |
4C4.9 + S100B/1012 | |
Unconjugated | |
XP_016883913 | |
Mouse | |
10-12kDa | |
Protein A/G Purified | |
RUO | |
6285 | |
IgG2a κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
S100B | |
Purified bovine brain S100 protein (4C4.9) ;Recombinant full-length human S100B protein (S100B/1012) | |
Brain, Melanoma | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |