Learn More
S100A5 Antibody (001), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Marca:  Novus Biologicals NBP2-90581AF350
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18215716
  • 607.98€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock
Descripción
Descripción
S100A5 Monoclonal specifically detects S100A5 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones
Especificaciones
S100A5
Monoclonal
Alexa Fluor 350
50mM Sodium Borate
Rabbit
Protein A purified
RUO
6276
Store at 4°C in the dark.
IgG
Immunohistochemistry, Immunohistochemistry (Paraffin)
1
Immunohistochemistry, Immunohistochemistry-Paraffin
Protein S-100D, S100 calcium binding protein A5, S100 calcium-binding protein A5S100Dprotein S100-A5
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse S100A5 (P63084; Met1-Lys93).
0.1 mL
Primary
Mouse
Purified