missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ S100A12 (Human) Recombinant Protein
Human S100A12 full-length ORF ( NP_005612.1, 1 a.a. - 92 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00006283-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MTKLEEHLEGIVNIFHQYSVRKGHFDTLSKGELKQLLTKELANTIKNIKDKAVIDEIFQGLDANQDEQVDFQEFISLVAIALKAAHYHTHKE
Especificaciones
NP_005612.1 | |
S100 calcium binding protein A12 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
S100A12 | |
GST |
6283 | |
Wheat germ expression system | |
10 μg | |
CAAF1, CAGC, CGRP, ENRAGE, MRP6, p6 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |