Learn More
Alfa Aesar™ (S)-3-Benzyloxycarbonylamino-2-(Boc-amino)propionic acid dicyclohexylammonium salt, 98%
Marca:  Alfa Aesar™ H61558.03
Detalles adicionales : CAS : 65710-58-9 Peso : 0.00100kg
Cantidad. 1g
 Ver más versiones de este producto
Código de producto. 15495515
  • 59.90€ / 1 g
Stock EU 4
Añadir a la cesta
Especificaciones
65710-58-9
519.683
RPWGTQRQPVPFKR-YDALLXLXSA-N
2755949
CC(C)(C)OC(=O)NC(CNC(=O)OCC1=CC=CC=C1)C(=O)O.C1CCC(CC1)NC2CCCCC2
C28H45N3O6
MFCD00236880
boc-dap z-oh dcha, a-z-l-2,3-diaminopropionic acid dicyclohexylammonium salt, dicyclohexylamine s-3-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino propanoate, boc-dap z-oh.dcha, boc-dap z-oh dicyclohexylammonium salt, pubchem23819, boc-dap z-oh?cha, boc-3-z-amino-l-alanine dicyclohexylammonium salt, boc-dap z-oh dicyclohexylammonium salt hplc, n, a-boc-n
N-cyclohexylcyclohexanamine;(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(phenylmethoxycarbonylamino)propanoic acid
Especificaciones
1g
65710-58-9
MFCD00236880
boc-dap z-oh dcha, a-z-l-2,3-diaminopropionic acid dicyclohexylammonium salt, dicyclohexylamine s-3-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino propanoate, boc-dap z-oh.dcha, boc-dap z-oh dicyclohexylammonium salt, pubchem23819, boc-dap z-oh?cha, boc-3-z-amino-l-alanine dicyclohexylammonium salt, boc-dap z-oh dicyclohexylammonium salt hplc, n, a-boc-n
CC(C)(C)OC(=O)NC(CNC(=O)OCC1=CC=CC=C1)C(=O)O.C1CCC(CC1)NC2CCCCC2
519.683
385.54
98%
(S)-3-Benzyloxycarbonylamino-2-(Boc-amino)propionic acid dicyclohexylammonium salt
C28H45N3O6
4100906
RPWGTQRQPVPFKR-YDALLXLXSA-N
N-cyclohexylcyclohexanamine;(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(phenylmethoxycarbonylamino)propanoic acid
2755949
Solid
Videos
SDS Documentos