missing translation for 'onlineSavingsMsg'
Learn More

(S)-1,3-Dibencilpiperazina, 97 %, Thermo Scientific™

Catalog Number p-7035036
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
204327-96-8
C18H22N2
266.388
MFCD03787922
AJNDRRBUHVZNKU-SFHVURJKSA-N
s-1,3-dibenzylpiperazine, 3s-1,3-dibenzylpiperazine, s-n4-benzyl-2-benzylpiperazine, 1-benzyl-3 s-benzylpiperazine, piperazine,1,3-bis phenylmethyl-, 3s
10825673
(3S)-1,3-dibencilpiperazina
C1CN(CC(N1)CC2=CC=CC=C2)CC3=CC=CC=C3
Este artículo no se puede devolver. Vea la política de devoluciones
204327-96-8
C18H22N2
266.388
MFCD03787922
AJNDRRBUHVZNKU-SFHVURJKSA-N
s-1,3-dibenzylpiperazine, 3s-1,3-dibenzylpiperazine, s-n4-benzyl-2-benzylpiperazine, 1-benzyl-3 s-benzylpiperazine, piperazine,1,3-bis phenylmethyl-, 3s
10825673
(3S)-1,3-dibencilpiperazina
C1CN(CC(N1)CC2=CC=CC=C2)CC3=CC=CC=C3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material (S)-1,3-Dibenzylpiperazine
Cantidad 250 mg
Formula Weight (peso de la fórmula) 266.39
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'