missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RWDD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17036-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The Formin-like protein 1 (FRL1) gene encodes a formin-related protein which has been implicated in morphogenesis, cyokinesis, and cell polarity. Formins are a conserved class of proteins expressed in all eukaryotes and have one DAD (diaphanous autoregulatory domain), one FH2 (formin homology 2) domain and one GBD/FH3 (Rho GTPase-binding / formin homology 3) domain. FRL1 is located in the cytoplasm and is highly expressed in the spleen, lymph node and bone marrow cells. FRL1 possibly has a role in the control of cell motility, survival of macrophages and cytoskeletal organization.Especificaciones
RWDD1 | |
Polyclonal | |
Unconjugated | |
RWDD1 | |
CGI 24, DFRP2, PTD013, RWD domain containing 1, RWDD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
259218, 51389, 66521 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99ND9, Q9CQK7, Q9H446 | |
RWDD1 | |
RWDD1 Fusion Protein Ag10525 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |