Learn More
Fisherbrand™ Rubber Belts for Van de Graaff Generator
Product Code. 12413120 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Width (Metric):
3.2 cm
Height (Metric):
81 cm long
Unit Size:
Each
Product Code. Width (Metric) Height (Metric) Unit Size
12413120 3.2 cm 81 cm long Each
1 options
This item is not returnable. View return policy
Product Code. 12413120
Quantity Price
1 21.87€ / paquete de 1
10 19.03€ / paquete de 1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Fisherbrand™ Rubber Belts for Van de Graaff Generator are well-suited for electrostatic generation.
  • Moving belt helps to accumulate electric charge
  • For use in electrostatic generation
Specifications
For Use With (Equipment) Junior Van
Height (Metric) 81 cm long
Product Type Rubber Belt for Van de Graaff Generator
Material Rubber
Width (Metric) 3.2 cm