missing translation for 'onlineSavingsMsg'
Learn More

RTN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11168-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16818063

  • 319.00€ / 150 microlitros

Envío estimado: 27-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene belongs to the family of reticulon encoding genes. Reticulons are associated with the endoplasmic reticulum, and are involved in neuroendocrine secretion or in membrane trafficking in neuroendocrine cells. Alternatively spliced transcript variants encoding different isoforms have been identified.
Especificaciones

Especificaciones

RTN2
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O70622, O75298
NSP like protein 1, NSP like protein I, NSP2, NSPL1, NSPLI, reticulon 2, RTN2
Rabbit
IgG
150 μL
-20°C
Primary
20167, 308410, 6253
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RTN2
Liquid
RTN2
RTN2 Fusion Protein Ag1655
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documentos
Certificados