Learn More
RPS6KA6 (RSK4) Recombinant Human Protein
RPS6KA6 (RSK4) Recombinant Human Protein
Marca:  Invitrogen PV4557
Detalles adicionales : Peso : 0.50000kg
Código de producto. 10516773
  • 632.00€ / 10 microgramos
Fecha estimada de envío: 16-04-2024
para ver el stock
Descripción
Descripción
RPS6KA6 (RSK4) is a member of the RSK kinases family. RSK family members are activated by ERKs and mitogens and are unusual in that they contain 2 non-identical kinase domains.
Especificaciones
Especificaciones
NP_055311
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay
RPS6KA6 (RSK4) Recombinant Human Protein
–80°C
1 Each
RPS6KA6 (RSK4) Recombinant Human Protein
RSK (RPS6K)
1016nmoles of phosphate transferred to PAKtide peptide substrate (CRRKSLVGpTPYWMAPE) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 1.7μg/mL
GST Tag
Kinase and inhibitors
90% by SDS-PAGE
0.44mg/mL
111.3kDa
10μg
For research use only. Not for use in diagnostic procedures
RPS6KA6 (RSK4)
RPS6KA6 (RSK4) was pre-diluted in enzyme dilution buffer and assayed in 25mM HEPES (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL PAKtide peptide substrate (CRRKSLVGpTPYWMAPE) and trace [32P]-γ-ATP for 10 minutes at 30°C
RPS6KA6 (RSK4) Recombinant Human Protein
Human
Baculovirus
Serine/threonine kinase
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Ofertas especiales
Ofertas especiales
For Research Use Only. Not for use in diagnostic procedures.