missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS6KA5 (MSK1) Recombinant Human Protein
RPS6KA5 (MSK1) Recombinant Human Protein
Marca: Invitrogen PV3681
Detalles adicionales : Peso : 0.05250kg
Descripción
RPS6KA5 (MSK1) is a serine⁄threonine protein kinase that is essential for the regulation of various cellular processes.Especificaciones
NP_004746.2 | |
Cell Viability, Pharma and Biopharma, Cell Proliferation and Function, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
119kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
RPS6KA5 (MSK1) | |
RPS6KA5 (MSK1) was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 100μM crosstide peptide substrate (GRPRTSSFAEG) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
RPS6KA5 (MSK1) Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.44mg/mL | |
9252 | |
RPS6KA5 (MSK1) Recombinant Human Protein | |
–80°C | |
1 Each | |
RPS6KA5 (MSK1) Recombinant Human Protein | |
RSK (RPS6K) | |
230nmoles of phosphate transferred to Crosstide peptide substrate (GRPRTSSFAEG) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 3.33μg/mL | |
GST Tag | |
Kinase and inhibitors | |
90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.