missing translation for 'onlineSavingsMsg'
Learn More

RPL39L Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 24940-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16813465

  • 319.00€ / 150 microlitros

Envío estimado: 22-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron containing gene within their coding regions. Ribosomal Protein L39 (RPL39), also known as 60S Ribosomal Protein L39, consists of 51 amino acids and is the smallest protein found in liver ribosomes. Ribosomal Protein L39 localizes to cytoplasm and is a member of the S39E family of ribosomal proteins. The gene encoding Ribosomal Protein L39 maps to human chromosome Xq24, and shares 92 percent homology with Ribosomal Protein L39L (Ribosomal Protein L39-like), which is also known as RPL39L, L39-2 or RPL39L1. Ribosomal Protein L39L is associated with the 60S ribosomal subunit and is expressed in testis with localization in nucleolus.
Especificaciones

Especificaciones

RPL39L
0.36 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96EH5
60S ribosomal protein L39 2, 60S ribosomal protein L39 like, ribosomal protein L39 like, RPL39L, RPL39L1
Rabbit
IgG
150 μL
-20°C
Primary
116832
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RPL39L
Liquid
RPL39L
RPL39L Fusion Protein Ag20923
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documentos
Certificados