missing translation for 'onlineSavingsMsg'
Learn More

RPL14 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP1-85385

Detalles adicionales : Peso : 0.00970kg

 Ver más versiones de este producto

Código de producto. 18215966

  • 484.87€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

RPL14 Polyclonal specifically detects RPL14 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

RPL14
Polyclonal
Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:1000-1:2500
CAG-ISL 7, CAG-ISL-7, CTG-B33, hRL14, L14, MGC88594, MMRPL32,60S ribosomal protein L14, ribosomal protein L14, RL14
Rabbit
Affinity Purified
RUO
9045
Human
IgG
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
RPL14
This antibody was developed against Recombinant Protein corresponding to amino acids:KKMRNRIIKNEVKKLQKAALLKASPKKAPGTKGTAAAAAAAAAAAAAAAKVPAKKITAASKKAPAQKVPAQKATGQKAAPAPKAQKGQKAPAQKAPAPKAS
0.1 mL
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only