Learn More
Fisherbrand™ Safety Coated Clear Straight Sided Round Bottles with White Polypropylene Pulp/Vinyl Cap
Click to view available options
Capacidad (métrico):
240 mL
480 mL
Cantidad:
12/Cs.
Descripción
Includes
ClosureColor,ClosureMaterial,LinerMaterial,dimColor,dimMaterial,Mouth,Shape,ClosureIncluded,dimSterility,dimType
Reduce breakage and lab mishaps with these Safety Coated Clear Straight Sided Round Bottles. If the glass breaks, the plastisol coating contains the substance in the bottle long enough to allow for proper disposal.
- Plastisol coating contains substance in bottle long enough to allow for proper disposal if the glass breaks
- Straight sided walls allow for the complete removal of contents
- Features polypropylene polypropylene pulp/vinyl cap
Información de órdenes
ClosureColor,ClosureMaterial,LinerMaterial,dimColor,dimMaterial,Mouth,Shape,ClosureIncluded,dimSterility,dimType
Especificaciones
Capacidad | 8 oz., 250mL |
Revestimiento del tapón | Polyvinyl |
Material del cierre | Polypropylene |
Tamaño del tapón | 70-400 mm |
Tipo de tapón | Screw Cap |
Tipo de producto | Safety-Coated Straight Sided Round Bottle |
Capacidad (métrico) | 240 mL |
Capacidad (imperial) | 8 oz. |
Color | Clear |
Material | Safety-coated Glass, Plastic-coated Glass |