Learn More
Gilac™ Round Dustbin with Black Handles
Product Code. 17958711
Change view
Click to view available options
Capacity (Metric):
50 L
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
17958711 50 L Each
1 options
This item is not returnable. View return policy
Product Code. 17958711 Supplier Gilac™ Supplier No. G0306211
  • ONLINE EXCLUSIVE PRICE
    97.68€
    64.63€ / paquete de 1
    Ahorro 33.05€
    33% Descuento
Fecha estimada de envoi: 07-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Round Dustbin with Black Handles is a round 50 L food container with handles.
  • Round 50 L dustbin with handles.
  • Lid available as an option.
Specifications
Capacity (Metric) 50 L
Capacity (English) 13.2 gal.
Color White
Autoclavable Not Autoclavable
Material Polyethylene
Product Type Dust Bin