Learn More
Gardner Denver Welch™ Rotary Evaporator Connection Kit
Product Code. 13514920 Shop All Welch Ilmvac Products
Change view
Click to view available options
For Use With (Equipment):
Chemical duty diaphragm pumps
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
13514920 Chemical duty diaphragm pumps Each
1 options
This item is not returnable. View return policy
Product Code. 13514920 Supplier Gardner Denver Welch™ Supplier No. 112575
  • ONLINE EXCLUSIVE PRICE
    141.00€
    118.60€ / paquete de 1
    Ahorro 22.40€
    15% Descuento
View all promotions
Fecha estimada de envoi: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Used for chemical duty diaphragm pumps.
  • Easy connection of the pump to common rotary evaporators
  • Includes vacuum hose 18/8x5 (1.5m), cooling water, chiller connection hose and clamps
TRUSTED_SUSTAINABILITY
Specifications
Type Rotary Evaporator Connection Kit
For Use With (Equipment) Chemical duty diaphragm pumps