missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROCK2(C-term) Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20248-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody is specific to ROCK2.
The function of this protein remains unknown.Especificaciones
ROCK2(C-term) | |
Polyclonal | |
Unconjugated | |
ROCK2 | |
KIAA0619, p164 ROCK 2, Rho kinase 2, ROCK2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19878, 25537, 9475 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75116, P70336, Q62868 | |
ROCK2 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |