Learn More
Cole-Parmer™ Rochester Pean Forceps
Product Code. 15458434 Shop All Cole-Parmer Products
Change view
Click to view available options
Length (Metric):
159 mm
Unit Size:
Each
Product Code. Length (Metric) Unit Size
15458434 159 mm Each
1 options
Product Code. 15458434 Supplier Cole-Parmer™ Supplier No. 1081812
  • ONLINE EXCLUSIVE PRICE
    32.90€
    23.46€ / paquete de 1
    Ahorro 9.44€
    28% Descuento
View all promotions
Fecha estimada de envoi: 22-02-2027
Add to basket
Add to basket
Manufactured from stainless steel. Cole-Parmer™ Rochester Pean Forceps includes a locking mechanism for clamping.
  • Standard grade
  • Straight jaw
  • Stainless steel
  • Length: 158.75mm
Specifications
Material Stainless Steel
Grade Standard
Tip Style Straight
Length (Metric) 159 mm
Not intended for medical applications or patient use.