missing translation for 'onlineSavingsMsg'
Learn More

RNPEPL1 Antibody (11G1), Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-59373

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215305

  • 611.22€ / 100 microgramos
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

RNPEPL1 Monoclonal specifically detects RNPEPL1 in Non-species specific samples. It is validated for Western Blot.
Especificaciones

Especificaciones

RNPEPL1
Monoclonal
Unconjugated
PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide
RNPEPL1
Synthetic N-terminal arginylated peptide conjugated to KLH
100 μg
Primary
Specific for N-terminal arginine, next to both glutamic acid and aspartic acid. Does not detect internal arginine.
Store at -20C.
IgG3
Western Blot
11G1
Western Blot 1:100 - 1:2000
argininyl aminopeptidase-like 1, arginyl aminopeptidase (aminopeptidase B)-like 1, arginyl aminopeptidase-like 1, EC 3.4.11.-, FLJ10806, FLJ26675, MGC99544, RNPEP-like 1
Mouse
Protein G purified
RUO
57140
Non-species specific
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only