missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF167 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24618-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
May act as an E3 ubiquitin-protein ligase, or as part of the E3 complex, which accepts ubiquitin from specific E2 ubiquitin-conjugating enzymes, such as UBE2E1, and then transfers it to substrates, such as SLC22A18. May play a role in growth regulation involved in G1/S transition.Especificaciones
RNF167 | |
Polyclonal | |
Unconjugated | |
RNF167 | |
5730408C10Rik, LP2254, ring finger protein 167, RING105, RNF167 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26001, 70510 | |
-20°C | |
Liquid |
Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q91XF4, Q9H6Y7 | |
RNF167 | |
RNF167 Fusion Protein Ag18383 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |