missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF135 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25061-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by RNF135 contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. This gene is located in a chromosomal region known to be frequently deleted in patients with neurofibromatosis.Especificaciones
RNF135 | |
Polyclonal | |
Unconjugated | |
RNF135 | |
L13, REUL, RIG I E3 ubiquitin ligase, ring finger protein 135, Riplet, RNF135 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
71956, 84282 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8IUD6, Q9CWS1 | |
Rnf135 | |
RNF135 Fusion Protein Ag18821 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |