Learn More
Applied Biosystems™ RNaseP Verification Plate, 96-well
Product Code. 15798669 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 x 96-well plate
Unit Size:
Each
Product Code. Quantity Unit Size
15798669 1 x 96-well plate Each
1 options
This item is not returnable. View return policy
Product Code. 15798669 Supplier Applied Biosystems™ Supplier No. 4432593
 Restricted Item
Fecha estimada de envoi: 30-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The 96-Well RNaseP Verification Plate is used to verify your real-time PCR instrument with Standard 96-Well Block
  • The 96-Well RNaseP Verification Plate is used to verify your real-time PCR instrument with Standard 96-Well Block.
  • Confirm the performance of your instrument by running the RNaseP assay. Please see your instrument user guide for full instructions on use.
  • The 96-Well RNaseP Verification Plate is preloaded with the necessary reagents to detect and quantitate genomic copies of the human RNase P gene, a single-copy gene encoding the RNA moiety of the RNase P enzyme.
  • Each well contains preloaded reaction mix (1X TaqMan™ Universal PCR Master Mix, RNase P primers, and FAM™ dye-labeled probe) and template.
  • The verification plate must demonstrate the ability to distinguish between 5,000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well.
  • To ensure accurate results, it is recommended that the performance of your instrument be verified every six months.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at –20 ± 5°C.
Calibration Type Instrument Verification Plate
Label or Dye Proprietary Mix
Shipping Condition Dry Ice
For Use With (Equipment) ViiA 7 Dx, QuantStudio Dx, QuantStudio 5 Dx, QuantStudio 7 Pro Dx
Quantity 1 x 96-well plate
For In Vitro Diagnostic Use