Learn More
Emma Safety Footwear Ringo Safety Shoes
Product Code. 15948931
Change view
Click to view available options
Size:
48
49
Unit Size:
1 pair
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15948931 49 1 pair
15938931 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15948931 Supplier Emma Safety Footwear Supplier No. MM73054049
  • ONLINE EXCLUSIVE PRICE
    129.72€
    76.39€ / 1 pares
    Ahorro 53.33€
    41% Descuento
Fecha estimada de envoi: 16-03-2027
Add to basket
Add to basket
This item is not returnable. View return policy
A versatile shoe with wipe clean uppers and an 'easy clean' sole. Popular with people working in Food and Catering. For peace of mind, an internal toe cap provides resilient protection against falling objects and unforeseen hazards.
  • S2-Safety shoe for wet working conditions
  • Full grain leather construction
  • GripForce™ Easy Twist Outsole
  • Hydro-Tec™ Sanitized Silver
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Black
Design Ringo
Liner Material Hydro-Tec™ Sanitized Silver
Material Leather
Size 49
Toe Material Steel