missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIC3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 27776-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Promotes functional expression of homomeric alpha-7 and alpha-8 nicotinic acetylcholine receptors at the cell surface. May also promote functional expression of homomeric serotoninergic 5- HT3 receptors, and of heteromeric acetylcholine receptors alpha- 3/beta-2, alpha-3/beta-4, alpha-4/beta-2 and alpha-4/beta-4.Especificaciones
RIC3 | |
Polyclonal | |
Unconjugated | |
RIC3 | |
AYST720, PRO1385, Protein RIC 3, RIC3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
320360, 79608 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q7Z5B4, Q8BPM6 | |
Ric3 | |
RIC3 Fusion Protein Ag26740 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |