missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHBDL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12467-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
RHBDL2 is a member of the rhomboid family of integral membrane proteins. This family contains proteins that are related to Drosophila rhomboid protein. Members of this family are found in both prokaryotes and eukaryotes and are thought to function as intramembrane serine proteases. RHBDL2 is thought to release soluble growth factors by proteolytic cleavage of certain membrane-bound substrates, including ephrin B2 and ephrin B3.Especificaciones
RHBDL2 | |
Polyclonal | |
Unconjugated | |
RHBDL2 | |
RHBDL2, Rhomboid like protein 2, Rhomboid related protein 2, RRP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
230726, 298512, 54933 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A2AGA4, Q9NX52 | |
RHBDL2 | |
RHBDL2 Fusion Protein Ag3142 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |