missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGN/SMP30 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17947-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a highly conserved, calcium-binding protein, that is preferentially expressed in the liver and kidney. It may have an important role in calcium homeostasis. Studies in rat indicate that this protein may also play a role in aging, as it shows age-associated down-regulation. This gene is part of a gene cluster on chromosome Xp11.3-Xp11.23. Alternative splicing results in two transcript variants having different 5′ UTRs, but encoding the same protein.Especificaciones
RGN/SMP30 | |
Polyclonal | |
Unconjugated | |
RGN | |
RC, Regucalcin, RGN, Senescence marker protein 30, SMP 30, SMP30 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19733, 25106, 9104 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q03336, Q15493, Q64374 | |
RGN | |
RGN/SMP30 Fusion Protein Ag12423 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |