Learn More
Fisherbrand™ Replacement Deuterium Lamp for Agilent UV and Visible HPLC Detector
Product Code. 15192738
Change view
Click to view available options
For Use With (Equipment):
1100 DAD, 8453 HPLC Detector
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15192738 1100 DAD, 8453 HPLC Detector Each
1 options
This item is not returnable. View return policy
Product Code. 15192738 Supplier Fisherbrand™ Supplier No. 1194112
  • ONLINE EXCLUSIVE PRICE
    910.00€
    821.11€ / paquete de 1
    Ahorro 88.89€
    9% Descuento
View all promotions
Fecha estimada de envoi: 29-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Replacement Deuterium Lamp for Agilent UV and Visible HPLC Detector has 2000 hours or 12 months shelf life.
  • Long life
  • Drop-in OEM replacement
  • Traceable by serial number
Compliance
WEEE and ROHS compliant
Specifications
Light Source Deuterium
Weight (English) 0.36 lb.
Weight (Metric) 0.16 kg
Description Item 14-500-721 is comparable to Agilent part number 2140-0605.
For Use With (Equipment) 1100 DAD, 8453 HPLC Detector
Product Type Detector Lamp