Learn More
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Sterile
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Sterile provide precise, positive-displacement dispensing of viscous or volatile liquids.
Marca:  Corning™ 6630
Detalles adicionales : Peso : 5.00000kg
 Ver más versiones de este producto
Código de producto. 15843905
  • 78.00€ / paquete de 100
Fecha de envío estimada
del proveedor 13-03-2019
Añadir a la cesta
Descripción
Descripción
  • Polypropylene tips for use with the Corning™ Step-R™ Repeating Pipettor
  • Available in eight sizes from 0.05mL to 50mL
  • Tips use the direct displacement method, dedicated for use with viscous, infectious and volatile liquids
  • Reliable and repeatable results through automated, individual tests of every single tip
  • Certification of every batch is available on request
Especificación
Especificación
Sterile
Dispensing of viscous and volatile liquids
Yes
100/Pk
Polypropylene
1.25mL
ISO8655 ISO11137
Videos