Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H
Product Code. 13265579
Change view
Click to view available options
Description:
Polyvalent, Phase 2
Polyvalent, Phases 1 and 2
Rapid Diagnostic 1 (b, d, E, r)
Rapid Diagnostic 2 (b, E, k, L)
Rapid Diagnostic 3 (d, E, G, k)
Salmonella H (1, 2)
Salmonella H (1, 5)
Salmonella H (1, 6)
Salmonella H (1, 7)
Salmonella H (E) Polyvalent
Salmonella H (L) Polyvalent
Salmonella H (a)
Salmonella H (b)
Salmonella H (c )
Salmonella H (d)
Salmonella H (eh)
Salmonella H (enx)
Salmonella H (gm)
Salmonella H (i)
Salmonella H (r )
Salmonella H (y)
Salmonella H (z27)
Unit Size:
2mL
Each
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
13265579 Polyvalent, Phase 2 2mL
13245579 Salmonella H (i) 2mL
11933342 Salmonella H (c ) 2mL
11983332 Salmonella H (a) 2mL
12697429 Salmonella H (1, 7) Each
12719290 Salmonella H (enx) Each
11923342 Salmonella H (b) 2mL
12789280 Salmonella H (d) Each
12667429 Salmonella H (1, 2) Each
12677429 Salmonella H (1, 5) Each
12687429 Salmonella H (1, 6) Each
12759290 Salmonella H (gm) Each
12677499 Polyvalent, Phases 1 and 2 Each
12759300 Salmonella H (r ) Each
12709290 Salmonella H (eh) Each
12617429 Salmonella H (z27) Each
12749280 Rapid Diagnostic 3 (d, E, G, k) Each
12719300 Salmonella H (L) Polyvalent Each
13235579 Salmonella H (E) Polyvalent Each
12660008 Salmonella H (y) Each
12697579 Rapid Diagnostic 1 (b, d, E, r) Each
12623658 Rapid Diagnostic 2 (b, E, k, L) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
This item is not returnable. View return policy
Product Code. 13265579 Supplier Thermo Scientific™ Supplier No. R30163201
  • ONLINE EXCLUSIVE PRICE
    153.00€
    114.54€ / 2 ml
    Ahorro 38.46€
    25% Descuento
 Restricted Item
Fecha estimada de envoi: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
  • Select sera also for use as positive controls for stained Salmonella suspensions
TRUSTED_SUSTAINABILITY
Specifications
Control Sets Positive
Description Polyvalent, Phase 2
Detectable Analytes Salmonella H Antigen
Type Agglutinating Sera
Material Polyvalent
Packaging Type 2 mL/Vial
Quantity 2 mL
Target Organism Class Salmonella H