Learn More
Masterflex™ Reducer Fitting, Barb x Barb, PVDF
Masterflex™ Reducer Fitting, Barb x Barb, PVDF are chemically resistant and ideal for low-pressure applications.
Marca:  Masterflex™ 30703-62
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 12479620
  • 28.00€ / paquete de 10
Fecha estimada de envío
28-08-2020
Añadir a la cesta
Descripción
Descripción
  • Ideal for low-pressure applications.
  • Quick and easy assembly without tools.
  • Chemically resistant.
  • Available in a variety of sizes, materials, and configurations.
  • Economical
  • Straight, barbed fittings that easily connect fluid paths using flexible tubing.
Especificaciones
Especificaciones
Barbed fitting, reducing connector
Reducing Connector with Barbed Fittings
Fits 5/8 in. ID x 1/2 in. ID Tubing
21 mm
Autoclavable
Fits 5/8 in. ID x 1/2 in. ID Tubing
Autoclave or Ethylene Oxide