missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Yeast GLRX2 Yeast Protein
A cDNA sequence encoding the GLRX2 Yeast was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11991-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
852124 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
METNFSFDSN LIVIIIITLF ATRIIAKRFL STPKMVSQET VAHVKDLIGQ KEVFVAAKTY CPYCKATLST LFQELNVPKS KALVLELDEM SNGSEIQDAL EEISGQKTVP NVYINGKHIG GNSDLETLKK NGKLAEILKP VFQ | |
Purity of GRX2 is greater than 90% as determined by SDS-PAGE. |
GLRX2 Protein | |
Research Use Only | |
GRX2 | |
Yeast | |
His | |
Glutaredoxin-2 solution contains 25mM Tris-HCl pH 7.5 and 0.01% Na Azide. |