missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Virus Parvovirus B19 VLP VP1 Protein

A cDNA sequence encoding the Parvovirus B19 VLP VP1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12966-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15922099

  • 183.00€ / 100 microgramos


Fecha estimada de envío
25-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

Virus Parvovirus B19 VLP VP1 Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Virus
Untagged
Greater than 90.0% as determined by 12% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
Parvovirus B19 VLP VP1 protein solution is supplied in PBS pH 7.4 and 0.02% Sodium azide.
Videos