missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Varicella Zoster Virus VZV ORF26 Protein
A cDNA sequence encoding the VZV ORF26 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13951-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Varicella Zoster Virus VZV ORF26 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris-Hcl pH 8.0, 1mM EDTA and 50% glycerol. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Varicella Zoster Virus | |
Untagged | |
Varicella protein is >95% pure as determined by 10% PAGE (coomassie staining). |