missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Toxoplasma Gondii Toxoplasma P22 Protein

A cDNA sequence encoding the Toxoplasma P22 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13787-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15914709

  • 691.00€ / 500 microgramos


Fecha estimada de envío
23-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

Toxoplasma Gondii Toxoplasma P22 Protein
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Toxoplasma Gondii
His
Toxoplasma protein is >95% pure as determined by 12% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
Sterile Filtered solution containing Phosphate buffered saline.
Videos