missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Toxoplasma Gondii T. gondii ROP4 Protein
A cDNA sequence encoding the T. gondii ROP4 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13668-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Toxoplasma Gondii T. gondii ROP4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris-HCl pH 8.0, 5mM Glutathione, 1.5M urea and 50% glycerol. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Toxoplasma Gondii | |
Untagged | |
Protein is >95% pure as determined by 10% PAGE (coomassie staining). |