missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae SPT15 Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) SPT15, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6764-ye-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
S. cerevisiae SPT15 Protein | |
Research Use Only | |
SPT15 | |
Recombinant Protein | |
Yeast | |
ADEERLKEFK EANKIVFDPN TRQVWENQNR DGTKPATTFQ SEEDIKRAAP ESEKDTSATS GIVPTLQNIV ATVTLGCRLD LKTVALHARN AEYNPKRFAA VIMRIREPKT TALIFASGKM VVTGAKSEDD SKLASRKYAR IIQKIGFAAK FTDFKIQNIV GSCDVKFPIR LEGLAFSHGT FSSYEPELFP GLIYRMVKPK IVLLIFVSGK IVLTGAKQRE EIYQAFEAIY PVLSEFRKM | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |